missing translation for 'onlineSavingsMsg'
Learn More

trans-3,5-Difluorocinnamic Acid 98.0+%, TCI America™

Numéro de catalogue. D41565G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D41565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. D41565G Fournisseur TCI America Code fournisseur D41565G
Il en reste null

Identifiants chimiques

CAS 147700-58-1
Molecular Formula C9H6F2O2
Molecular Weight (g/mol) 184.142
MDL Number MFCD00010321
InChI Key MBAWRXICVNIUGY-OWOJBTEDSA-N
Synonym 3,5-difluorocinnamic acid, trans-3,5-difluorocinnamic acid, 3,5-difluorocinnamicacid, e-3-3,5-difluorophenyl acrylic acid, 3-3,5-difluorophenyl acrylic acid, 2e-3-3,5-difluorophenyl prop-2-enoic acid, 2e-3-3,5-difluorophenyl acrylic acid, rarechem bk hw 0255, timtec-bb sbb006675, 3-3,5-difluorophenyl propenoic acid
PubChem CID 5374941
IUPAC Name (E)-3-(3,5-difluorophenyl)prop-2-enoic acid
SMILES C1=C(C=C(C=C1F)F)C=CC(=O)O

Spécifications

Melting Point 204°C
Color White
Quantity 5 g
Formula Weight 184.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-3,5-Difluorocinnamic Acid