missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Pentenoic acid, 90+%, remainder other isomers

Catalog No. AAA1268918
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1268918 50 g
AAA1268909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1268918 Supplier Thermo Scientific Chemicals Supplier No. A1268918

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13991-37-2 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13991-37-2
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00002704
InChI Key YIYBQIKDCADOSF-ONEGZZNKSA-N
Synonym trans-2-pentenoic acid, 2-pentenoic acid, e-pent-2-enoic acid, pent-2-enoic acid, 2e-pent-2-enoic acid, 2-pentenoic acid, 2e, beta-ethyl acrylic acid, c2h5ch=chcooh, pentenoic acid, unii-1rg66883cf
PubChem CID 638122
ChEBI CHEBI:38366
IUPAC Name (E)-pent-2-enoic acid
SMILES CC\C=C\C(O)=O

Specifications

Melting Point 8°C to 10°C
Density 0.992
Boiling Point 200°C to 203°C
Flash Point 102°C (215°F)
Odor Pungent
Refractive Index 1.4515
Quantity 50 g
UN Number UN3265
Beilstein 1720312
Formula Weight 100.12
Percent Purity ≥90%
Assay remainder other isomers
Chemical Name or Material trans-2-Pentenoic acid
Show More Show Less
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 237-791-9
RTECSNumber SB2750000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only