missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Methyl-2-butenal 95.0+%, TCI America™

Catalog No. T10035ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10035ML 5 mL
T100325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10035ML Supplier TCI America Supplier No. T10035ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 497-03-0
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00006977
InChI Key ACWQBUSCFPJUPN-HWKANZROSA-N
Synonym tiglic aldehyde, trans-2-methyl-2-butenal, tiglaldehyde, 2-methylbut-2-enal, e-2-methylbut-2-enal, tiglic acid aldehyde, e-2-methyl-2-butenal, 2-butenal, 2-methyl-, e, tigaldehyde, trans, 2-butenal, 2-methyl-, 2e
PubChem CID 5321950
IUPAC Name (E)-2-methylbut-2-enal
SMILES CC=C(C)C=O

Specifications

Color Yellow
Boiling Point 116°C
Quantity 5 mL
UN Number 1989
Formula Weight 84.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Methyl-2-butenal