missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Methyl-2-butenal, 97%

Catalog No. AAL0811314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0811314 25 g
AAL0811306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0811314 Supplier Thermo Scientific Chemicals Supplier No. L0811314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 497-03-0 | C5H8O | 84.118 g/mol

Tiglic aldehyde was used in the concise total synthesis of 7-demethylpiericidin A11. It was used as starting reagent during the synthesis of alkyl-branched tetraene hydrocarbons, pheromone component for dried fruit beetle.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tiglic aldehyde was used in the concise total synthesis of 7-demethylpiericidin A11. It was used as starting reagent during the synthesis of alkyl-branched tetraene hydrocarbons, pheromone component for dried fruit beetle.

Solubility
Soluble in water

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 497-03-0
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00006977
InChI Key ACWQBUSCFPJUPN-HWKANZROSA-N
Synonym tiglic aldehyde, trans-2-methyl-2-butenal, tiglaldehyde, 2-methylbut-2-enal, e-2-methylbut-2-enal, tiglic acid aldehyde, e-2-methyl-2-butenal, 2-butenal, 2-methyl-, e, tigaldehyde, trans, 2-butenal, 2-methyl-, 2e
PubChem CID 5321950
IUPAC Name (E)-2-methylbut-2-enal
SMILES CC=C(C)C=O

Specifications

Density 0.871
Boiling Point 117°C to 119°C
Flash Point 18°C (64°F)
Odor Fruit-like
Refractive Index 1.448
Quantity 25 g
UN Number UN1989
Beilstein 1698207
Sensitivity Air sensitive
Solubility Information Soluble in water
Formula Weight 84.12
Percent Purity 97%
Chemical Name or Material trans-2-Methyl-2-butenal
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALDEHYDES, N.O.S.
EINECSNumber 207-833-0
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only