missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexen-1-ol, 97%

Catalog No. AAA1327214
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1327214 25 g
AAA1327236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1327214 Supplier Thermo Scientific Chemicals Supplier No. A1327214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 928-95-0 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 928-95-0
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00002927
InChI Key ZCHHRLHTBGRGOT-SNAWJCMRSA-N
Synonym trans-2-hexen-1-ol, trans-2-hexenol, 2-hexenol, e-hex-2-en-1-ol, e-2-hexen-1-ol, 2-hexen-1-ol, 2-hexen-1-ol, 2e, 3-propylallyl alcohol, 2-hexen-1-ol, e, 2-hexen-1-ol, trans
PubChem CID 5318042
SMILES CCC\C=C\CO

Specifications

Density 0.843
Boiling Point 158°C to 160°C
Flash Point 54°C (129°F)
Odor Pleasant
Refractive Index 1.438
Quantity 25 g
UN Number UN1987
Beilstein 1719709
IUPAC Name (2E)-hex-2-en-1-ol
Formula Weight 100.16
Percent Purity 97%
Chemical Name or Material trans-2-Hexen-1-ol
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H319
Flammable liquid and vapour.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 213-191-2
RTECSNumber MP8390000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only