missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hepten-1-ol 96.0+%, TCI America™

Catalog No. H06825ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06825ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H06825ML Supplier TCI America Supplier No. H06825ML

Chemical Identifiers

CAS 33467-76-4
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00014056
InChI Key ASFYPVGAALGVNR-AATRIKPKSA-N
Synonym trans-2-hepten-1-ol, e-2-hepten-1-ol, e-hept-2-en-1-ol, 2-hepten-1-ol, e, 2-hepten-1-ol, 2e, 2-heptenol, unii-q989yfo10i, 2-hepten-1-ol, hept-2-en-1-ol, 2e-hept-2-en-1-ol
PubChem CID 5318017
IUPAC Name (E)-hept-2-en-1-ol
SMILES CCCCC=CCO

Specifications

Color Colorless
Boiling Point 75°C
Quantity 5 mL
UN Number 1987
Formula Weight 114.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Hepten-1-ol
EINECSNumber (2)-2393
TSCA Yes