missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Aminocyclohexanol hydrochloride, 99%, Thermo Scientific™

Numéro de catalogue. p-3777237
Modifier l'affichage
Click to view available options
Quantité:
5g
25g
Emballage:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC206980050 5g Glass bottle
AC206980250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC206980050 Fournisseur Thermo Scientific Chemicals Code fournisseur 206980050

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 5456-63-3
Formule moléculaire C6H13ClNO
Poids moléculaire (g/mol) 150.63
Numéro MDL MFCD00001490,MFCD09259962,MFCD09259962,MFCD09259963,MFCD11618002,MFCD11618003,MFCD09259963
Clé InChI SEBSRZIHRZRNAH-KGZKBUQUSA-N
Synonyme trans-2-aminocyclohexanol hydrochloride, 1r,2r-2-aminocyclohexanol hydrochloride, 1r, 2r-2-aminocyclohexanol hydrochloride, 1r,2r-2-aminocyclohexanol hcl, 1r,2r-trans-2-aminocyclohexanol hydrochloride, trans-2-hydroxycyclohexylamine hydrochloride, 1r,2r-2-aminocyclohexan-1-ol hydrochloride, cyclohexanol, 2-amino-, hydrochloride, 1r,2r-rel, 1r,2r-2-aminocyclohexanolhydrochloride, 1r,2r-rel-2-aminocyclohexanol hydrochloride
PubChem CID 12228412
Nom de l’IUPAC (1R,2R)-2-aminocyclohexan-1-ol;hydrochloride
SOURIRES [Cl].N[C@@H]1CCCC[C@H]1O

Spécifications

Nom ou substance chimique trans-2-Aminocyclohexanol hydrochloride
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 99%
Emballage Glass bottle
Formule linéaire H2NC6H10OH·HCl
Quantité 5g
Beilstein 13, III, 704
Renseignements sur la solubilité Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,ether,benzene
Poids de formule 151.64
Pourcentage de pureté 99%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only