missing translation for 'onlineSavingsMsg'
Learn More

trans-1,4-Cyclohexanediamine 98.0+%, TCI America™

Catalog No. C14265G
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14265G 5 g
C142625G 25 g
C1426100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C14265G Supplier TCI America Supplier No. C14265G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2615-25-0
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00035862,MFCD00075174,MFCD08276323
InChI Key VKIRRGRTJUUZHS-UHFFFAOYSA-N
Synonym trans-1,4-diaminocyclohexane, trans-cyclohexane-1,4-diamine, 1,4-diaminocyclohexane, trans-1,4-cyclohexanediamine, cis-cyclohexane-1,4-diamine, 1,4-cyclohexanediamine, cis-1,4-cyclohexanediamine, 1,4-cyclohexanediamine, trans, 1r,4r-cyclohexane-1,4-diamine, 1,4-cyclohexanediamine, cis
PubChem CID 18374
IUPAC Name cyclohexane-1,4-diamine
SMILES NC1CCC(N)CC1

Specifications

Melting Point 71°C
Boiling Point 88°C
Quantity 5 g
UN Number 3259
Formula Weight 114.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-1,4-Cyclohexanediamine