missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Cyclohexanediol, 97%

Catalog No. AC161650500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161650500 50 g Glass bottle
AC161650100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC161650500 Supplier Thermo Scientific Chemicals Supplier No. 161650500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1460-57-7 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1460-57-7
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00063611
InChI Key PFURGBBHAOXLIO-PHDIDXHHSA-N
Synonym trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq & trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol
PubChem CID 92886
ChEBI CHEBI:16931
IUPAC Name (1R,2R)-cyclohexane-1,2-diol
SMILES O[C@@H]1CCCC[C@H]1O

Specifications

Melting Point 100°C to 104°C
Color White
Boiling Point 117°C (17.0 Mbar)
Flash Point 134°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H10(OH)2
Quantity 50 g
Fieser 17,91
Solubility Information Solubility in water: 375g/L (20 c)
Viscosity 8 mPa.s (110°C)
Formula Weight 116.16
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material trans-1, 2-Cyclohexanediol, 98%
Show More Show Less
EINECSNumber 215-956-6

RUO – Research Use Only