missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Quinine, MilliporeSigma™ Supelco™

Catalog No. 111012925
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012925 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012925 Supplier Merck Emd Millipore Supplier No. 6931150MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Specifications

CAS 130-95-0
Product Line TraceCERT
Physical Form Neat
Linear Formula C20H24N2O2
MDL Number MFCD00198096
Melting Point 173°C to 175°C
Synonym 6′-Methoxycinchonidine
Type Certified Reference Material
Quantity 50 mg
Formula Weight 324.42
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC