missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Phosphate Standard for IC, 1,000 mg/L Phosphate in Water, MilliporeSigma™ Supelco™

Catalog No. 111015439
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015439 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015439 Supplier Merck Emd Millipore Supplier No. 38364100ML

missing translation for 'validMainframeMsg'

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.Certified content incl. uncertainty and expiry date are given on the certificate.

Specifications

Product Line TraceCERT
CAS 7558-79-4
Linear Formula Na2 HPO4
MDL Number MFCD00003496
Synonym Sodium phosphate dibasic solution; Disodium hydrogen phosphate solution; di-Sodium hydrogen phosphate solution
Type Certified Reference Material
Quantity 100 mL
Formula Weight 141.96
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 1,000 mg/L phosphate aq. soln.

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC