missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Phenylbutazone certified reference material, MilliporeSigma™ Supelco™

Catalog No. 111019148
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019148 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019148 Supplier Merck Emd Millipore Supplier No. 80184100MG
Only null left
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Product Line TraceCERT
CAS 50-33-9
Physical Form Neat
Linear Formula C19H20N2O2
MDL Number MFCD00005500
Melting Point 106°C to 108°C
Synonym 4-Butyl-1,2-diphenyl-3,5-pyrazolidinedione
Type Certified Reference Material
Quantity 100 mg
Formula Weight 308.37
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC