missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Penconazol, Certified Reference Material, MilliporeSigma™ Supelco™
Spécifications
Spécifications
| CAS | 66246-88-6 |
| Flash Point | 100°C (Closed Cup) |
| Molecular Formula | C13H15Cl2N3 |
| Linear Formula | C13H15Cl2N3 |
| Quantity | 50 mg |
| MDL Number | MFCD00078737 |
| Formula Weight | 284.18 |
| Grade | Trace CERT |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC