missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Nicotinic acid, MilliporeSigma™ Supelco™

Catalog No. 111001968
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001968 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001968 Supplier Merck Emd Millipore Supplier No. 7983850MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nicotinic acid is a water soluble B complex vitamin, with effective plasma lipid-lowering effect and hence is clinically used to treat hyperlipidemia.

Vitamins, Nutraceuticals and Natural Products

Specifications

Product Line TraceCERT
CAS 59-67-6
Flash Point 193°C - Closed Cup
Physical Form Powder
Linear Formula C6H5 NO2
MDL Number MFCD00006391
Melting Point 236°C to 239°C
Synonym 3-Picolinic acid; Niacin; Pellagra preventive factor; Pyridine-3-carboxylic acid; Vitamin B3
Type Certified Reference Material
Quantity 50 mg
Formula Weight 123.11
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC