missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Mono-Methyl fumarate, MilliporeSigma™ Supelco™

Catalog No. 111013799
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013799 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013799 Supplier Merck Emd Millipore Supplier No. 9257150MG
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Specifications

CAS 2756-87-8
Product Line TraceCERT
Linear Formula HO2 CCH=CHCO2 CH3
MDL Number MFCD00063174
Melting Point 144°C to 145°C
Synonym Fumaric acid monomethyl ester; Methyl hydrogen fumarate
Type Certified Reference Material
Quantity 50 mg
Formula Weight 130.10
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC