missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Lactate Standard for IC, 1,000 mg/L Lactate in Water, MilliporeSigma™ Supelco™

Catalog No. 111004790
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004790 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004790 Supplier Merck Emd Millipore Supplier No. 07096100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.Certified content incl. uncertainty and expiry date are given on the certificate.

Specifications

Product Line TraceCERT
Purity Grade Notes 1000 mg/L lactate aq. soln., prepared with magnesium L-lactate hydrate and water; filtered through a 0.2 mum membrane filter and stabilized with sodium azide
Concentration 1000 mg/L lactate aq. soln.
Type Certified Reference Material
Quantity 100 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC