missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Iprodione certified reference material, MilliporeSigma™ Supelco™
Specifications
Specifications
| Product Line | TraceCERT |
| CAS | 36734-19-7 |
| Linear Formula | Cl2C6H3N2C3H2O2 CONHCH(CH3)2 |
| MDL Number | MFCD00055353 |
| Melting Point | 130°C to 134°C |
| Synonym | [3-(3,5-Dichlorophenyl)-2,4-dioxoimidazolidinyl]-N-(methylethyl)carboxamide; Iprodione |
| Type | Certified Reference Material |
| Quantity | 50 mg |
| Formula Weight | 330.17 |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC