missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Fludioxonil certified reference material, MilliporeSigma™ Supelco™

Numéro de catalogue. 111018441
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111018441 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111018441 Fournisseur Merck Emd Millipore Code fournisseur 7412350MG
Il en reste null

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Spécifications

Product Line TraceCERT
CAS 131341-86-1
Linear Formula C12H6F2N2O2
MDL Number MFCD01631158
Type Certified Reference Material
Quantity 50 mg
Formula Weight 248.19
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC