missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Ethyl p-Toluenesulfonate, MilliporeSigma™ Supelco™

Catalog No. 111015592
Change view
Click to view available options
Quantity:
120 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015592 120 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015592 Supplier Merck Emd Millipore Supplier No. 68383120MG
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Specifications

CAS 80-40-0
Product Line TraceCERT
Density 1.174 g/mL (at 25°C)
Flash Point 158°C - closed cup
Linear Formula CH3C6H4 SO3C2H5
MDL Number MFCD00009100
Melting Point 29°C to 33°C
Boiling Point 158°C to 162°C (10 mmHg)
Type Certified Reference Material
Quantity 120 mg
Formula Weight 200.25
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC