missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ EPA 8260 Internal Standards Mix, 2,000 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111017316
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017316 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017316 Supplier Merck Emd Millipore Supplier No. CRM48958

Specifications

Packaging Ampule
CAS 67-56-1
Product Line TraceCERT
Flash Point 9.7°C - closed cup
Concentration 2,000 μg/mL each component in methanol
Synonym EPA 8260 A Internal Standards Mix
Type Certified Reference Material
Quantity 1 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC