missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Dimethyl Terephthalate Solution, 5 mg/g in DMSO-d, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111022478
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022478 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022478 Supplier Merck Emd Millipore Supplier No. 393875X1ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI. Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

Specifications

CAS 120-61-6
Flash Point 87°C (Closed Cup)
Molecular Formula C10H10O4
Linear Formula C10H10O4
Quantity 5 x 1 mL
MDL Number MFCD00008440
Formula Weight 194.18
Grade Trace CERT™ 1 H-qNMR Standard
Concentration or Composition (by Analyte or Components) 5 mg/g in DMSO-d6

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC