missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Di-n-butyl phthalate Solution, 5,000 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111020105
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020105 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020105 Supplier Merck Emd Millipore Supplier No. 40066
Only null left

Di-n-butyl phthalates (DnBPs) are mainly used in personal-care products like perfumes, aftershaves, shampoos, make up and nail products, paints, etc.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 84-74-2
Flash Point 9°C
Type Food and Cosmetic Component Standard
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 5,000 μg/mL in methanol

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC