missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Cobalt Standard for ICP, 1,000 mg/L Co in Nitric Acid, MilliporeSigma™ Supelco™

Catalog No. 111020327
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020327 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020327 Supplier Merck Emd Millipore Supplier No. 30329100MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Product Line TraceCERT
CAS 64-17-5
Type Certified Reference Material
Quantity 100 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 1,000 mg/L Co in nitric acid

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC