missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Aroclor Solution 1260, 1 000 μg/mL en isooctane, MilliporeSigma™ Supelco™

Numéro de catalogue. 111020313
Modifier l'affichage
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111020313 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111020313 Fournisseur Merck Emd Millipore Code fournisseur 44809
Environnement

Spécifications

Packaging Ampoule
Product Line TraceCERT
CAS 11096-82-5
Flash Point -12°C
Type Organic Pollutant Standard
Quantity 1 mL
Grade TraceCERT™ Norme analytique
Concentration or Composition (by Analyte or Components) 1,000 μg/mL in isooctane

TraceCERT est une marque déposée de Sigma-Aldrich Co. LLC