missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Aroclor 1242 Solution, 1,000 μg/mL in isooctane, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| CAS | 53469-21-9 |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 1,000 μg/mL in isooctane |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC