missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Aldrin certified reference material, MilliporeSigma™ Supelco™

Numéro de catalogue. 111018524
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111018524 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111018524 Fournisseur Merck Emd Millipore Code fournisseur 0857350MG
Il en reste null

Spécifications

Product Line TraceCERT
CAS 309-00-2
Linear Formula C12H8 Cl6
MDL Number MFCD00135587
Type Certified Reference Material
Quantity 50 mg
Formula Weight 364.91
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC