missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ 52 Component Indoor Air Standard, 100 μg/mL each component in Methanol, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| CAS | 67-56-1 |
| Synonym | Japanese Indoor Air Standard Mix |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 100 μg/mL each component in methanol: water (95:5) |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC