missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ 2,4,6-Trichlorophenol Solution, 5,000 μg/mL in Methanol, MilliporeSigma™ Supelco™

Numéro de catalogue. 111020110
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111020110 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111020110 Fournisseur Merck Emd Millipore Code fournisseur 40019
Il en reste null
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Spécifications

Packaging Ampule
Product Line TraceCERT
CAS 88-06-2
Flash Point 11°C
Type Food and Cosmetic Component Standard
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 5,000 μg/mL in methanol

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC