Learn More
Fisher Education™ Traceable™ Precision Monitoring Thermohygrometer with Calibration
Simultaneously display min/max and current temperature and humidity with 7-day graph
Supplier:  Fisher Education™ 0800047
Catalog No. S94141
Only null left
Description
Description
This digital temperature and humidity meter continuously monitors and displays readings numerically and graphically. A week's worth of data is kept safe and the memory cannot be cleared, only overwritten by the next day's measurements.
  • The internal sensor makes these ideal for use in hoods, stores, cleanrooms, incubators, and drying or environmental chambers.
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Scroll through temperature and humidity data for the past seven days
  • Maintain data integrity—historical data cannot be cleared, only overwritten
  • Easily switch between fahrenheit and celsius and 12/24-hour time formats
  • Traceable™ model number 6418
  • Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
  • Guaranteed to have at least one year of calibration remaining on certificate
  • 1 year warranty
Specifications
Specifications
Monitoring Thermohygrometer
0.1% RH
±0.4°C
0 to 90%
-20°C to +50°C
7 x 1 x 4 in.
Product Suggestions
Product Suggestions