missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Memory-Loc™ Ultra-Low Temperature USB Datalogging Refrigerator/Freezer Thermometer

Catalog No. 15079623
Change view
Click to view available options
Model:
Memory-Loc, 1 probe
Memory-Loc, 2 probes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
15079623 Memory-Loc, 2 probes
15079622 Memory-Loc, 1 probe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15079623 Supplier Fisherbrand™ Supplier No. 9367986

Traceable™ Datalogging Thermometer that can be used in 21 CFR 11 environments

Fisher Scientific™ Traceable™ Memory-Loc™ Datalogging Thermometers can be used in 21 CFR 11 environments. High-accuracy thermometer has a temperature range of -90°C to 105°C (-130F to 221°F)and resolution of 0.01 and maintains an accuracy of ±0.25°C across the entire temperature range. Ideal for monitoring temperatures in refrigerators, freezers, water baths, heating blocks, and incubators.
  • Data-logging thermometer with fixed one-minute timing intervals
  • Meets current CDC requirements and additional recommendations for vaccine storage and monitoring
  • Can be used in 21 CFR 11 environments, raw data is locked and cannot be cleared or changed on base unit
  • Stores up to ten unique alarm events
  • Capable of storing over 1 million temperature observations
  • Hassle-free retrieval of data; unit can remain in use while downloading and analyzing data
  • Status indicators: Low battery, low memory, USB data transfer and active alarm state
  • Traceable to NIST for accuracy
  • High-impact, chemical-resistant ABS plastic case measures 7 x 10.8 x 1.9cm (2.75 x 4.25 x 0.75 in.)
  • Weight: 5 oz.
Hi/Lo Alarms and Time/Date Stamps
  • Alarm tracking feature stores in memory each alarm event detailing when the alarm state occurred, as well as when the unit returned back to within range
  • Alarm state indicators include visual LEDs, audio alerts, and flashing LCD segments
  • Data for up to ten different alarm events can be retrieved with the most current event viewable on the display
USB Data Transferring
  • All new feature allows recorded data (CSV file) to be transferred from thermometer to PC (or Mac™) using a USB flash drive (not included)
  • No software is needed for computer interface
  • Each thermometer has a unique ID allowing multiple units to be used in the same location
  • Raw data on unit cannot be overwritten or modified
  • Indicator light will appear when 95% of memory is full
Data-Logging Features
  • Can be used in environments requiring 21 CFR 11 compliance
  • Thermometer logs two years of observations with a one-minute logging interval
  • Device will stop recording when storage is full and will not overwrite recorded data
  • Data is unable to be deleted from memory once stored
Bottle Probe Features
  • Bottle probes are sealed in a miniature bottle (1 x 2.5 in.) filled with nontoxic glycol
  • Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration)
  • Eliminates concerns about incidental contact with food or drinking water
  • Temperature-buffered bottle sensor eliminates rapid changes when refrigerator door is opened
  • Ideal for use in monitoring storage conditions for vaccines or other important samples
  • Meets all current CDC requirements and recommendations for temperature monitoring of vaccine storage as part of the Vaccines for Children program
  • Temperature Range: -90°C to 105°C (-130F to 221°F)
  • Temperature Resolution: 0.01°
  • Accuracy: ±0.2°C
  • Calibration Points: -80°C, -40°C, 0°C and 100°C
  • Part Number 15-079-622 includes 1 316 Stainless Steel Probe with Platinum RTD Sensor
  • Part Number 15-079-623 includes 2 316 Stainless Steel Probes with Platinum RTD Sensors

Specifications

Type Temperature Datalogger
Display Type LCD
Calibration Calibrated
Alarm Types LED Indicator, Audible, Flashing LCD Segment
Software Type TraceableLIVE
Interface USB
Model Memory-Loc, 2 probes
Sensor Types Platinum RTD, Temperature-Buffered Bottle Sensor
Accuracy (Temperature) ±0.2°C
Range -90°C to +105°C (-130°F to +221°F)
Temperature Resolution 0.01°
Temperature Range (English) -130°F to 221°F
Temperature Range (Metric) -90°C to +105°C
Show More Show Less