missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Memory-Loc™ Datalogging Thermometer

Catalog No. 15081130
Change view
Click to view available options
Includes:
1 Bottle
1 Probe, 5/16 in. dia. x 3/4 in. long
1 Stainless-steel Probe
1 Vaccine Bottle Probe
2 Bottles
2 Probes (Bottle Probe and 5/16 in. dia. x 3/4 in. long)
2 Probes, 5/16 in. dia. x 3/4 in. long
2 Stainless-steel Probes
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
15081130 1 Bottle
15081134 2 Probes (Bottle Probe and 5/16 in. dia. x 3/4 in. long)
15081136 2 Stainless-steel Probes
15081132 1 Probe, 5/16 in. dia. x 3/4 in. long
15081131 2 Bottles
15081135 1 Stainless-steel Probe
15081133 2 Probes, 5/16 in. dia. x 3/4 in. long
15079619 1 Vaccine Bottle Probe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 15081130

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Traceable™ Memory-Loc™ Datalogging Thermometer that can be used in 21 CFR 11 environments.

  • Data-logging thermometer with fixed one-minute timing intervals
  • Meets current CDC requirements and additional recommendations for vaccine storage and monitoring
  • Can be used in 21 CFR 11 environments, raw data is locked and cannot be cleared or changed on base unit
  • Stores up to ten unique alarm events
  • Capable of storing over 1 million temperature observations
  • Hassle-free retrieval of data; unit can remain in use while downloading and analyzing data
  • Status indicators: Low battery, low memory, USB data transfer and active alarm state
  • Traceable to NIST for accuracy
  • High-impact, chemical-resistant ABS plastic case measures 7 x 10.8 x 1.9 cm (2.75 x 4.25 x 0.75 in.)
  • Weight: 5 oz.

Hi/Lo Alarms and Time/Date Stamps

  • Alarm tracking feature stores in memory each alarm event detailing when the alarm state occurred, as well as when the unit returned back to within range
  • Alarm state indicators include visual LEDs, audio alerts, and flashing LCD segments
  • Data for up to ten different alarm events can be retrieved with the most current event viewable on the display

USB Data Transferring

  • All new feature allows recorded data (CSV file) to be transferred from thermometer to PC (or Mac™) using a USB flash drive (not included)
  • No software is needed for computer interface
  • Each thermometer has a unique ID allowing multiple units to be used in the same location
  • Raw data on unit cannot be overwritten or modified
  • Indicator light will appear when 95% of memory is full

Data-Logging Features

  • Can be used in environments requiring 21 CFR 11 compliance
  • Thermometer logs two years of observations with a one-minute logging interval
  • Device will stop recording when storage is full and will not overwrite recorded data
  • Data is unable to be deleted from memory once stored

Bottle Probe Features

  • Bottle probes are sealed in a miniature bottle (1 x 2.5 in.) filled with nontoxic glycol
  • Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration)
  • Eliminates concerns about incidental contact with food or drinking water
  • Temperature-buffered bottle sensor eliminates rapid changes when refrigerator door is opened
  • Ideal for use in monitoring storage conditions for vaccines or other important samples
  • Meets all current CDC requirements and recommendations for temperature monitoring of vaccine storage as part of the Vaccines for Children program

Order Info

Replacement battery: Cat. No. 1111

Specifications

Type Datalogger
Display Type LCD
Calibration NIST
Alarm Types LED Indicator, Audible, Flashing LCD Segment
Software Type TraceableLIVE
Description 1 Bottle
Includes 1 Bottle
Certifications/Compliance NIST, ISO/IEC 17025, A2LA, CDC
Interface USB
Sensor Types Temperature-Buffered Bottle Sensor
Accuracy (Temperature) ±0.25°C
Temperature Resolution 0.01°
Temperature Range (Metric) -50°C to +70°C
Operating Temperature (English) -58°F to +158°F
Operating Temperature (Metric) -72.4°F to +316.4°F
Weight (English) 5 oz.
Weight (Metric) 141.7 g
Show More Show Less