missing translation for 'onlineSavingsMsg'
Learn More

Trace Metals AA Sample 2, MilliporeSigma™ Supelco™

Catalog No. 111016994
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016994 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016994 Supplier Merck Emd Millipore Supplier No. QC127320ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 7697-37-2
Type Certified Reference Material
Quantity 20 mL
Grade Certified Reference Material