missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TPPP3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA524925

Numéro de catalogue. PIPA524925


Il en reste null

Description

Description

This antibody is predicted to react with bovine and rat based on sequence homology.

TPPP3 binds tubulin and has microtubule bundling activity. TPPP3 may play a role in cell proliferation and mitosis.
Spécifications

Identifiants chimiques

TPPP3
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human, Mouse, Rat
Tppp3
2700055K07Rik; brain specific protein; Ceacam9; CGI-38; hypothetical protein LOC614988; mmCGM8; p20; p25gamma; RGD1305061; TPPP/p20; TPPP3; tubulin polymerization promoting protein family member 3; tubulin polymerization-promoting protein family member 3
Tppp3
Primary
Protein A, Antigen affinity chromatography
0.5 mg/mL
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
RUO
PBS with 0.09% sodium azide; pH 7.4
Q5PPN5, Q9BW30, Q9CRB6
291966, 51673, 67971
KLH conjugated synthetic peptide between 117-146 amino acids of human TPPP3.
Antibody

Spécifications

TPPP3
Polyclonal
Unconjugated
Tppp3
2700055K07Rik; brain specific protein; Ceacam9; CGI-38; hypothetical protein LOC614988; mmCGM8; p20; p25gamma; RGD1305061; TPPP/p20; TPPP3; tubulin polymerization promoting protein family member 3; tubulin polymerization-promoting protein family member 3
Rabbit
Protein A, Antigen affinity chromatography
RUO
291966, 51673, 67971
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q5PPN5, Q9BW30, Q9CRB6
Tppp3
KLH conjugated synthetic peptide between 117-146 amino acids of human TPPP3.
400 μL
Primary
Human, Mouse, Rat
Antibody
IgG
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions