missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ BLaER1 Human B-cell Precursor Leukemia Cell Line

Catalog No. SCC165
Change view
Click to view available options
Quantity:
≥ 1 x 106 cells/vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCC165 ≥ 1 x 106 cells/vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC165 Supplier MilliporeSigma™ Supplier No. SCC165

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BLaER1 human B-cell precursor leukemia cell line may be transdifferentiated and used to model human monocytes.

  • BLaER1 is a highly efficient model for transdifferentiation of B-cell-derived human cancer cells. Each vial contains ≥1 x 10 viable cells.
  • The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.
  • Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
  • Cells are verified to be of human origin and negative for inter-species contamination from mouse, rat, chinese hamster, Golden Syrian hamster, and non-human primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services Cells are negative for mycoplasma contamination.
  • Cells tested positive for squirrel monkey retrovirus (SMRV).

Specifications

Content And Storage Store in liquid nitrogen
Cell Type Cancer Cells
Culture Type Mammalian Cell Culture
Form Frozen
Gender Female
Species Human
Tissue Bone Marrow
Format Vial
Quantity ≥ 1 x 106 cells/vial