missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toxicity Characteristic Leachate Procedure Standard 1, SPEX CertiPrep™
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ba | 500μg/mL |
| Ag | 25μg/mL |
| As | 25μg/mL |
| Cr | 25μg/mL |
| Pb | 25μg/mL |
| Cd | 5μg/mL |
| Se | 5μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | TCLP Standard 1 without Mercury |
| Packaging | Triple-leached LDPE bottle |
| Name Note | Without Mercury |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| CAS | 7440-39-3, 7440-22-4, 7440-38-2, 7697-37-2, 7732-18-5 |
| Analyte or Component Names | Ba( 500mg/L),Ag(25mg/L),As(25mg/L) |
| Technique | AA, ICP |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Element | Ba, Ag, As |
| Show More |
Product intended for laboratory use only