Learn More
Brady™ ToughStripe™ Die-Cut Floor Marking Number 3
Catalog No. 19108324
Change view
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19108324 Yellow
19108319 Black
19108320 White
19108321 Red
19108322 Green
19108323 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19108324 Supplier Brady™ Supplier No. 121870
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For marking aisle-ways, shipping and receiving docks, distribution, or other areas of your facility
  • Developed for superior durability
  • Easy application
  • Painless removal
  • More visible than paint
Specifications
Adhesive Type Permanent Rubber-based
Color Yellow