Learn More
Brady™ ToughStripe™ Die-Cut Floor Marking Letter X
Description
- Visible floor-marking improves safety and organization in almost any industrial environment
- High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
- Tape removes cleanly without any unsightly adhesive residue or damage to the floor
- Exceeds safety standards for clean, dry non-slip surfaces per ANSI A 1264.2-2006 and OSHA 1910.22
Spécifications
Spécifications
| Adhesive Type | Permanent Rubber-based |
| Color | Black |