missing translation for 'onlineSavingsMsg'
Learn More

Total Recoverable Petroleum Hydrocarbons (TRPH) - WP, MilliporeSigma™ Supelco™

Catalog No. 111017077
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017077 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017077 Supplier Merck Emd Millipore Supplier No. QC12722ML
Only null left

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

Packaging Ampule
CAS 76-13-1
Type Certified Reference Material
Quantity 2 mL
Grade Certified Reference Material