missing translation for 'onlineSavingsMsg'
Learn More

Total Phenolics, MilliporeSigma™ Supelco™

Catalog No. 111017061
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017061 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017061 Supplier Merck Emd Millipore Supplier No. QC11342ML
Only null left

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 67-64-1
Flash Point -17°C - closed cup - Solvent
Type Certified Reference Material
Quantity 2 mL
Grade Certified Reference Material