Learn More
RPC Group E-Z Chek Sensitive Total Chlorine Test Strips, 0.1 ppm, 100/bottle
Supplier: RPC Group K1000106
E-Z Chek Sensitive Total Chlorine & Chloramines Test Strips provide a quick and convenient means for indicating low levels of total chlorine in water. Monitoring between carbon tanks for chlorine/chloramines break through is the primary application for the E-Z Chek Sensitive.