missing translation for 'onlineSavingsMsg'
Learn More

Toluene, 99+%, extra pure

Numéro de catalogue. AC177160025
Modifier l'affichage
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC177160025 2.5 L Glass Bottle
AC177160010 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC177160025 Fournisseur Thermo Scientific Chemicals Code fournisseur 177160025
Il en reste null

CAS: 108-88-3 | C7H8 | 92.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Spécifications

Melting Point -95.0°C
Density 0.8660g/mL
Quantity 2.5 L
Flash Point 4°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4968
Residue after Evaporation 0.003% max.
Assay Percent Range 99+%
Specific Gravity 0.866
Linear Formula C6H5CH3
Vapor Pressure Vapor Pressure: 29mbar at 20°C
Viscosity 0.6 mPa.s (20°C)
Water 0.05% max. (K.F.)
Beilstein 05, 280
Merck Index 15, 9688
Solubility Information Solubility in water: almost insoluble (0.5g/L at 20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, acetone,, ether, petroleum ether, benzene, ch2cl2, dmf, gla, cial acetic acid, chcl3, ccl4.
Boiling Point 111.0°C
Formula Weight 92.14
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Toluene
Afficher plus de résultats Afficher moins de résultats
Hazard Category Signal Word: DANGER!
Hazard Statement GHS P Statement May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damag
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

RUO – Research Use Only