missing translation for 'onlineSavingsMsg'
Learn More

Toluene, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020617
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020617 5 mL
111019471 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020617 Supplier Merck Emd Millipore Supplier No. 896805ML

Toluene is an aromatic hydrocarbon, which can be obtained from crude oil via cracking or distillation process.

Featured Industries: Environmental

Specifications

CAS 108-88-3
Ignition Point 997°F
Vapor Pressure 26 mmHg (25°C)
Vapor Density 3.2 (vs. air)
Density 0.865 g/mL (at 25°C)
Flash Point 4.0°C
Physical Form Neat
Linear Formula C6H5 CH3
MDL Number MFCD00008512
Melting Point -93°C
Boiling Point 110°C to 111°C
Type Analytical Standard
Quantity 5 mL
Formula Weight 92.14
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less