missing translation for 'onlineSavingsMsg'
Learn More

Tofisopam 98.0+%, TCI America™

Catalog No. T350950MG
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T350950MG 50 mg
T3509250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T350950MG Supplier TCI America Supplier No. T350950MG

Chemical Identifiers

CAS 22345-47-7
Molecular Formula C22H26N2O4
Molecular Weight (g/mol) 382.46
MDL Number MFCD00823171
InChI Key RUJBDQSFYCKFAA-UHFFFAOYSA-N
Synonym 1-(3,4-Dimethoxyphenyl)-5-ethyl-7,8-dimethoxy-4-methyl-5H-benzo[d][1,2]diazepine, 1-(3,4-Dimethoxyphenyl)-5-ethyl-7,8-dimethoxy-4-methyl-5H-2,3-benzodiazepine
PubChem CID 5502
IUPAC Name 1-(3,4-dimethoxyphenyl)-5-ethyl-7,8-dimethoxy-4-methyl-5H-2,3-benzodiazepine
SMILES CCC1C(=NN=C(C2=CC(=C(C=C12)OC)OC)C3=CC(=C(C=C3)OC)OC)C

Specifications

Melting Point 157°C
Color White-Yellow
Quantity 50 mg
UN Number 3077
Formula Weight 382.46
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tofisopam