missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tofacitinib citrate

CAS: 540737-29-9 | C22H28N6O8, C16H20N6O·C6H8O7 | 504.50 g/mol

$318.81 - $1051.94

Chemical Identifiers

CAS 540737-29-9
Molecular Formula C16H20N6O·C6H8O7
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC466530010
View Documents
Thermo Scientific Chemicals
466530010
1 g
Each for $1,051.94
Only null left
 
AC466532500
View Documents
Thermo Scientific Chemicals
466532500
250 mg
Each for $318.81
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

540737-29-9
C16H20N6O·C6H8O7

Specifications

540737-29-9
Solubility aq. soln.: soluble. Other solubilities: soluble in dmso
C22H28N6O8
SYIKUFDOYJFGBQ-UHFFFAOYNA-N
2-hydroxypropane-1,2,3-tricarboxylic acid; 3-{4-methyl-3-[methyl({7H-pyrrolo[2,3-d]pyrimidin-4-yl})amino]piperidin-1-yl}-3-oxopropanenitrile
Tofacitinib citrate
1 g
198.0°C to 202.0°C
504.49
OC(=O)CC(O)(CC(O)=O)C(O)=O.CC1CCN(CC1N(C)C1=C2C=CNC2=NC=N1)C(=O)CC#N
504.50
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only