accessibility menu, dialog, popup

UserName

CAS RN 540737-29-9

IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid; 3-{4-methyl-3-[methyl({7H-pyrrolo[2,3-d]pyrimidin-4-yl})amino]piperidin-1-yl}-3-oxopropanenitrile
Synonyms: citric acid; tofacitinib
Molecular Weight (g/mol): 504.5
Molecular Formula: C22H28N6O8
InChi Key: SYIKUFDOYJFGBQ-UHFFFAOYNA-N
SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O.CC1CCN(CC1N(C)C1=C2C=CNC2=NC=N1)C(=O)CC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)