missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tofacitinib citrate

Catalog No. AC466530010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466530010 1 g
AC466532500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466530010 Supplier Thermo Scientific Chemicals Supplier No. 466530010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 540737-29-9 | C22H28N6O8, C16H20N6O·C6H8O7 | 504.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 540737-29-9
Molecular Formula C22H28N6O8

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: soluble. Other solubilities: soluble in dmso
Melting Point 198.0°C to 202.0°C
Formula Weight 504.49
InChI Key SYIKUFDOYJFGBQ-UHFFFAOYNA-N
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O.CC1CCN(CC1N(C)C1=C2C=CNC2=NC=N1)C(=O)CC#N
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid; 3-{4-methyl-3-[methyl({7H-pyrrolo[2,3-d]pyrimidin-4-yl})amino]piperidin-1-yl}-3-oxopropanenitrile
Molecular Weight (g/mol) 504.50
Chemical Name or Material Tofacitinib citrate

RUO – Research Use Only