missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Todd Hewitt Broth with Colistin and Nalidixic Acid (LIM Broth)

Catalog No. R064814
Change view
Click to view available options
Quantity:
100/Pk.
Fill Volume:
5 mL
Manufacturing Origin:
United States
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Fill Volume Manufacturing Origin
R064814 100/Pk. 5 mL United States
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R064814 Supplier Thermo Scientific™ Supplier No. R064814

Reduce media prep time while selectively isolating group B streptococci from clinical specimens containing mixed flora with pre-filled, ready-to-use tubes.

Selectively isolate group B streptococci from clinical specimens containing mixed flora using Thermo Scientific™ Todd Hewitt Broth with Colistin and Nalidixic Acid (LIM Broth).

Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes of Todd Hewitt Broth with the addition of Colistin and Nalidixic Acid to inhibit growth of gram-negative bacteria, available in multiple formats and formulations.

Streptococci are facultatively anaerobic, catalase-negative, gram-positive cocci that occur as pairs or chains. They cause a wide variety of clinical diseases including pharyngitis, scarlet fever, otitis media, impetigo, meningitis, neonatal sepis, pneumonia, endocarditis, osteomyelitis, and necrotizing fascitis. One of the most useful phenotypic characteristics of streptococci is the hemolytic reaction, generally classified as alpha, beta or gamma according to the appearance of zones around colonies growing on Blood Agar. Beta-hemolytic streptococci are further characterized by Lancefield groups based on the antigenic differences in group-specific polysaccharides located in the bacterial cell wall.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications

Description Todd Hewitt Broth (LIM Broth)
Format 15mm x 103mm Tube
Manufacturing Origin United States
Product Type Microbiology Media
Fill Volume 5 mL
Quantity 100/Pk.