missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112769
Description
TOB1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
This gene encodes a member of the tob/btg1 family of anti-proliferative proteins that have the potential to regulate cell growth. When exogenously expressed, this protein supresses cell growth in tissue culture. The protein undergoes phophorylation by a serine/threonine kinase, 90 kDa ribosomal S6 kinase. Interactions of this protein with the v-erb-b2 erythroblastic leukemia viral oncogene homolog 2 gene product p185 interferes with growth suppression. This protein inhibits T cell proliferation and transcription of cytokines and cyclins. The protein interacts with both mothers against decapentaplegic Drosophila homolog 2 and 4 to enhance their DNA binding activity. This interaction inhibits interleukin 2 transcription in T cells.Identifiants chimiques
| TOB1 | |
| -20°C or -80°C if preferred | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| TOB1 | |
| APRO5; APRO6; MGC104792; MGC34446; PIG49; proliferation-inducing gene 49; protein Tob1; Protein Tob1-like protein; TOB; TOB1; transducer of erbB2; Transducer of erbB-2 1; transducer of ERBB2, 1; transducer of ErbB-2.1; Trob; TROB1; Unknown (protein for MGC:134447) | |
| TOB1 | |
| Primary | |
| Protein G |
| 2.9 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P50616 | |
| 10140 | |
| Recombinant Human Protein Tob1 protein (42-175AA). | |
| Antibody |
Spécifications
| TOB1 | |
| Polyclonal | |
| Unconjugated | |
| TOB1 | |
| APRO5; APRO6; MGC104792; MGC34446; PIG49; proliferation-inducing gene 49; protein Tob1; Protein Tob1-like protein; TOB; TOB1; transducer of erbB2; Transducer of erbB-2 1; transducer of ERBB2, 1; transducer of ErbB-2.1; Trob; TROB1; Unknown (protein for MGC:134447) | |
| Rabbit | |
| Protein G | |
| RUO | |
| 10140 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.9 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P50616 | |
| TOB1 | |
| Recombinant Human Protein Tob1 protein (42-175AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |