missing translation for 'onlineSavingsMsg'
Learn More

TMS-PZ (=N-Trimethylsilylimidazole in Anhydrous Pyridine), TCI America™

Catalog No. T062312ML
Change view
Click to view available options
Quantity:
12 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T062312ML 12 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T062312ML Supplier TCI America Supplier No. T062312ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

[Trimethylsilylating Reagent, for wet or dry Sugars, Mono or Poly Saccharides]

Chemical Identifiers

MDL Number MFCD00148688

Specifications

Color Colorless
Quantity 12 mL
Physical Form Clear Liquid at 20°C
UN Number 2924
Chemical Name or Material TMS-PZ (=N-Trimethylsilylimidazole in Anhydrous Pyridine) [Trimethylsilylating Reagent, for w
TSCA No
Recommended Storage Refrigerator