missing translation for 'onlineSavingsMsg'
Learn More

TMS-HT (=HMDS and TMCS in Anhydrous Pyridine) [for Gas Chromatography], TCI America™

Numéro de catalogue. A56045ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A56045ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. A56045ML Fournisseur TCI America Code fournisseur A56045ML
Il en reste null

Identifiants chimiques

MDL Number MFCD00192427
Synonym Hexamethyldisilazane and Trimethylchlorosilane in Anhydrous Pyridine, Trimethylchlorosilane and Hexamethyldisilazane in Anhydrous Pyridine

Spécifications

Color Yellow
Quantity 5 mL
Physical Form Clear Liquid-Slightly Cloudy Liquid at 20°C
UN Number 2924
Chemical Name or Material TMS-HT (=HMDS and TMCS in Anhydrous Pyridine) [for Gas Chromatography]
TSCA No
Recommended Storage Refrigerator